Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Buckets for 4-Place Swinging Bucket Rotors
Catalog No. 75006446
Change view
Click to view available options
Capacity:
1 x 250 mL
1 x 750 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75006446 1 x 250 mL
75006441 1 x 750 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75006446 Supplier Thermo Scientific™ Supplier No. 75006446
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Round or rectangular buckets for use with Four-Place Swinging Bucket Rotors 75006445 and 75006434
Order Info
Sold individually; order correct quantity to equip rotor. Sealing lids sold separately. Accessory adapters accept assorted tube styles and volumes from 1.5 to 250mL.
Specifications
For Use With (Equipment) Swinging bucket rotor
Capacity 1 x 250 mL
Product Type Bucket
Description Rectangular Bucket