Learn More
Thermo Scientific™ Sorvall™ Legend™ Micro 17 Microcentrifuge PROMO
Accelerate routine sample preparation up to 17,000 x g with generous capacity in a compact microcentrifuge footprint.
Supplier:  Thermo Scientific™ 75002431PR
Catalog No. 75-002-431PM
Only null left
Explore available promotions
Description
Description
Support your micro-volume separation needs with:
  • Performance up to 17,000 x g with fast acceleration and deceleration.
  • Choose from 7 optional microcentrifuge rotors to find the ideal fit for your application:
    • Outstanding corrosion resistance with lightweight engineered polymer rotors.
    • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns.
    • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids.
    • Eliminate the need for adapters with unique dual row rotor.
  • Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure.
  • Sleek, lightweight, space-saving design.
  • Durable and easy to clean and maintain.
  • Quiet performance promotes a safe, stress-free environment.
  • Conforms to the latest clinical and safety standards, including 98/79/EC In Vitro Diagnostic Directive.
Specifications
Specifications
24 x 1.5/2 mL
17,000 x g
Ambient Temperature
Benchtop Centrifuge
120 V, 60 Hz
No
IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
23.1 lb.
50/60 Hz
1.5/2 mL Rotor with Clickseal Biocontainment Lid
13,300 rpm
56 dBA
CSA, CE, IVD
24 x 1.5/2.0 mL rotor with ClickSeal biocontainment lid
2 Years
1 to 99 min. (1 min. increments); HOLD
22.5 x 24.3 x 35.2 cm (8.9 x 9.6 x 13.8 in.)
Sorvall
120 V
Product Suggestions
Product Suggestions