missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan monooleate

Catalog No. AAJ62576K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62576K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62576K2 Supplier Thermo Scientific Chemicals Supplier No. J62576K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sorbitan monooleate, CAS # 1338-43-8, is a synthetic fatty acid ester showing emulsifying and dispersing activities. | CAS: 1338-43-8 | C24H44O6 | 428.61 g/mol

An emulsifier Sorbitan monooleate is used as a lipophillic non-ionic surfactant, emulsifier, stabilizer for oil based lotions, ointments, in textiles as antistat, lubricant for fabric softener, sweetening agent, food additive, toothpaste, tobacco, toiletries and in cosmetics. It serves as a dispersant in oil field chemicals and as a corrosion Inhibitor. Further, it plays as an important role as excipient and intravenous osmotic diuretic in pharmaceutical fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Sorbitan monooleate is derived from sorbitol and is a synthetic fatty acid ester that presents emulsifying, dispersing, and wetting activities

Application

• Sorbitan monooleate is used in yeast laboratory strains to protect them from excess drying and also helps rehydrate yeast cells

• It can be used as a dispersant and as a corrosion inhibitor

• Sorbitan monooleate has important uses as an excipient and intravenous osmotic diuretic

Chemical Identifiers

CAS 1338-43-8
Molecular Formula C24H44O6
Molecular Weight (g/mol) 428.61
MDL Number MFCD00080948
InChI Key NWGKJDSIEKMTRX-NZAQQJATSA-N
Synonym Span™ 80
PubChem CID 70693544
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Color Yellow
Physical Form Viscous Liquid
Quantity 1 L
Flash Point 113°C (235°F)
Merck Index 14,8724
Solubility Information Soluble in ethanol at 50mg/mlMiscible with water,ethanol,isopropanol and ether; Insoluble in acetone.
Formula Weight 428.61
Chemical Name or Material Sorbitan monooleate
EINECSNumber 215-665-4
RTECSNumber WG2932400
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only