missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan Monooleate, Spectrum™ Chemical

Catalog No. 18613969
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613969 500 mL Poly Drum
18613968 4 L Poly Drum
18613967 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613969 Supplier Spectrum Chemical Mfg Cor Supplier No. SPA80500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1338-43-8

Specifications

Acidity 8
CAS Max % 1
Packaging Poly Drum
Quantity 500 mL
Saponification Value 145 - 160
Molecular Weight (g/mol) 428.61
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Sorbitan Monooleate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.