missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan Monolaurate, Spectrum™ Chemical

Catalog No. p-7158298
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
20 L
200 L
4 L
500 mL
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Catalog No. Quantity Packaging
18606919 500 mL Amber Glass Bottle
18606920 4 L Amber Glass Bottle
18606921 20 L Poly Drum
18606922 200 L Drum
4 options

Catalog No. 18606919

Supplier: Spectrum Chemical Mfg Cor SPA20500MLGL

Only null left

SPA20, 1338-39-2, C18H34O6

Spectrum™ Chemical Sorbitan Monolaurate , also known as sorbitan laurate, is a mix of esters from the fatty acid lauric acid used as a food additive emulsifier to keep water and oils mixed. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 1338-39-2
Molecular Formula C18H34O6
Molecular Weight (g/mol) 346.46
MDL Number MFCD00005365
InChI Key LWZFANDGMFTDAV-IOVMHBDKSA-N
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Saponification Value 158-175
Grade Reagent