accessibility menu, dialog, popup

UserName

H3COOOH(R)O(R)OH(S)OH

CAS RN 1338-39-2

IUPAC Name: 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
Synonyms: sorbitan laurate
Molecular Weight (g/mol): 346.46
Molecular Formula: C18H34O6
InChi Key: LWZFANDGMFTDAV-IOVMHBDKSA-N
SMILES: CCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)