missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbic Acid 99.0+%, TCI America™
$36.57 - $86.64
Chemical Identifiers
| CAS | 110-44-1 |
|---|---|
| Molecular Formula | C6H8O2 |
| Molecular Weight (g/mol) | 112.13 |
| MDL Number | MFCD00002703 |
| InChI Key | WSWCOQWTEOXDQX-MQQKCMAXSA-N |
| Synonym | sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid |
| PubChem CID | 643460 |
| ChEBI | CHEBI:38358 |
| IUPAC Name | (2E,4E)-hexa-2,4-dienoic acid |
| SMILES | C\C=C\C=C\C(O)=O |
Chemical Identifiers
| 110-44-1 | |
| 112.13 | |
| WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
| 643460 | |
| (2E,4E)-hexa-2,4-dienoic acid |
| C6H8O2 | |
| MFCD00002703 | |
| sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid | |
| CHEBI:38358 | |
| C\C=C\C=C\C(O)=O |
Specifications
| 110-44-1 | |
| White | |
| C6H8O2 | |
| 25 g | |
| WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
| (2E,4E)-hexa-2,4-dienoic acid | |
| 643460 | |
| 112.13 | |
| Crystalline Powder |
| 134°C | |
| 228°C | |
| MFCD00002703 | |
| sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid | |
| C\C=C\C=C\C(O)=O | |
| 112.13 | |
| CHEBI:38358 | |
| ≥99.0% (HPLC,T) | |
| Sorbic Acid |
Safety and Handling
EINECSNumber : (2)-0609&(2)-1075
RTECSNumber : WG2100000
TSCA : Yes