missing translation for 'onlineSavingsMsg'
Learn More

Sorbic acid, free acid, ≥99%, MP Biomedicals™

$62.53 - $62.53

Chemical Identifiers

CAS 110-44-1
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00002703
InChI Key WSWCOQWTEOXDQX-MQQKCMAXSA-N
Synonym sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
PubChem CID 643460
ChEBI CHEBI:38358
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
SMILES C\C=C\C=C\C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10293780
View Documents
MP Biomedicals Inc
0210293780
100 g
Each for $62.53
Only null left
 
Description

Description

Specifications

Chemical Identifiers

110-44-1
112.13
WSWCOQWTEOXDQX-MQQKCMAXSA-N
643460
(2E,4E)-hexa-2,4-dienoic acid
C6H8O2
MFCD00002703
sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
CHEBI:38358
C\C=C\C=C\C(O)=O

Specifications

110-44-1
1.2g/cm3 at 20°C
MFCD00002703
sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
C\C=C\C=C\C(O)=O
112.13
CHEBI:38358
≥99%
Sorbic acid, free acid
Yellow
C6H8O2
100 g
WSWCOQWTEOXDQX-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienoic acid
643460
112.13
Crystalline Powder
Videos
SDS
Documents

Documents