missing translation for 'onlineSavingsMsg'
Learn More

Sorbic acid, 99%

Catalog No. aaa1619636
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1619636 500 g
AAA1619630 250 g
AAA161960B 1000 g
AAA161960I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1619636 Supplier Thermo Scientific Chemicals Supplier No. A1619636
Only null left

CAS: 110-44-1 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sorbic acid, is used as a food additive and has functional use such as antimicrobial preservatives, fungistatic agents. Sorbic acid is used as a preservative and is found in food, cosmetics, and other household products. It prevents the growth of mold, yeast, and fungi. It acts as a stabilizers and antioxidants.

Solubility
It is hardly soluble in water but soluble in glacial acetic acid, ethanol, propane diol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from Bases, Oxidizing agents, Reducing agentsBases, Oxidizing agents, Reducing agents. Photosensitive.

Chemical Identifiers

CAS 110-44-1
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00002703
InChI Key WSWCOQWTEOXDQX-MQQKCMAXSA-N
Synonym sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
PubChem CID 643460
ChEBI CHEBI:38358
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
SMILES C\C=C\C=C\C(O)=O

Specifications

Melting Point 132°C to 135°C
Density 1.205
Boiling Point 228°C (decomposition)
Flash Point 127°C (260°F)
Linear Formula CH3CH=CHCH=CHCO2H
Quantity 500 g
Beilstein 1741831
Merck Index 14,8721
Solubility Information It is hardly soluble in water but soluble in glacial acetic acid,ethanol,propane diol.
Formula Weight 112.13
Percent Purity 99%
Chemical Name or Material Sorbic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-768-7
RTECSNumber WG2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only