Learn More
SOMNI Scientific™ SOMNI Scientific™ SOMNI Ped Coaxial Circuit
The tube within a tube design eliminates twisting of the inspiratory and expiratory limbs and provides heat and moisture conservation. PVC, Phthalate and Latex Free
$106.73
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
View More Specs
Products 1
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
04777207
View Documents Promotion Details
SOMNI Scientific™
BC13003
60 in.
Each for $106.73
Only null left
Description
Description
  • A tube within a tube design that eliminates twisting of the inspiratory and expiratory limbs.
  • PVC, Phthalate and Latex Free
  • Provides heat and moisture conservation
Specifications
Specifications
Research Animal Inhalant Anesthesia