Learn More
SOMNI Scientific™ SOMNI Scientific™ SOMNI Ped Coaxial Circuit
Description
- A tube within a tube design that eliminates twisting of the inspiratory and expiratory limbs.
- PVC, Phthalate and Latex Free
- Provides heat and moisture conservation
Specifications
Specifications
| For Use With (Application) | Research Animal Inhalant Anesthesia |
| Length (English) | 60 in. |