missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solvent Yellow 124, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 34432-92-3 |
| Physical Form | Neat |
| Linear Formula | C22H31N3O2 |
| Synonym | N-Ethyl-N-[2-(1-isobutoxyethoxy)ethyl]-p-(phenylazo)aniline |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 369.50 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Analytical Standard |