missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles
GREENER_CHOICE

Catalog No. p-7161460
Change view
Click to view available options
Chemical Label:
Ethanol
Isopropanol
Methanol
Unlabeled
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Label
FB0340936 Ethanol
FB0340935 Isopropanol
FB0340933 Methanol
FB0340934 Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FB0340936 Supplier Fisherbrand™ Supplier No. FB0340936
Only null left

Pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.

  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available

Use LYOB only with solvents compatible with LDPE.

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Translucent
Material LDPE
Closure Color Natural
Certifications/Compliance OSHA 29 CFR 1910.1200
Mouth Wide
Chemical Label Ethanol
Closure Material Polypropylene
Closure Size 53 mm
Autoclavable Not Autoclavable
Show More Show Less