missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles

Description
- Wide mouth 2 in. (53mm) bottle for easy refilling
- Polypropylene closure with leakproof design, color-coded to aid in easy identification
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Use LYOB only with solvents compatible with LDPE.
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Translucent |
| Material | LDPE |
| Closure Color | Yellow |
| Mouth | Wide |
| Chemical Label | Isopropanol |
| Closure Material | Polypropylene |
| Closure Size | 53 mm |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 16 oz. |
| Show More |