missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Solvent Still Head
Grease free, easy syringe access, and easy, controlled, drainage.
Supplier: Synthware™ S244504
Description
Grease free solvent still head with a ground glass joint and a 2 mm T-Bore Stopcock on the delivery port. Also features 4 mm stopcocks on the upper portion for easy syringe needle access.
Specifications
| PTFE T-Bore Plug and Stopcock | |
| Solvent Still Head | |
| 500 mL | |
| Top: 24/40; Bottom: 24/40 |
| Organic Synthesis | |
| Borosilicate Glass | |
| 1 Ea. |