missing translation for 'onlineSavingsMsg'
Learn More

Solvent Red 197 98.0+%, TCI America™

Catalog No. p-7135989
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09191G 1 g
S09195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09191G Supplier TCI America Supplier No. S09191G
Only null left

Chemical Identifiers

CAS 52372-39-1
Molecular Formula C23H19N5O
Molecular Weight (g/mol) 381.44
MDL Number MFCD02176212
InChI Key FXFIDVQMNRVEGQ-UHFFFAOYSA-N
Synonym 3-(Diethylamino)-7-imino-7H-benzo[4,5]imidazo[1,2-a]chromeno[3,2-c]pyridine-6-carbonitrile
PubChem CID 104185
IUPAC Name 17-(diethylamino)-11-imino-14-oxa-3,10-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2,4,6,8,12,15,17,19-nonaene-12-carbonitrile
SMILES CCN(CC)C1=CC=C2C=C3C4=NC5=CC=CC=C5N4C(=N)C(C#N)=C3OC2=C1

Specifications

Formula Weight 381.44
Percent Purity ≥98.0% (HPLC,N)
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Solvent Red 197
EINECSNumber (5)-5365
TSCA Yes