accessibility menu, dialog, popup

UserName

CAS RN 52372-39-1

H3CNCH3NNHNNO

CAS RN 52372-39-1

IUPAC Name: 17-(diethylamino)-11-imino-14-oxa-3,10-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2,4,6,8,12,15,17,19-nonaene-12-carbonitrile
Synonyms: 17-(diethylamino)-11-imino-14-oxa-3,10-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2,4,6,8,12,15,17,19-nonaene-12-carbonitrile
Molecular Weight (g/mol): 381.44
Molecular Formula: C23H19N5O
InChi Key: FXFIDVQMNRVEGQ-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=C3C4=NC5=CC=CC=C5N4C(=N)C(C#N)=C3OC2=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results