missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Solvent Blue 38

CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol

Supplier:  Thermo Scientific Chemicals A1539522

 View more versions of this product

Catalog No. AAA1539522

Only null left
Explore available promotions

Description

Description

Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.

Solubility
Soluble in water (partly), and methanol (1 mg/ml).

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Solvent Blue 38
1328-51-4
MFCD00071424
Soluble in water (partly),and methanol (1mg/ml).
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2]
92030797
100 g
∼235°C (decomposition)
C32H14CuN8Na2O6S2
C.I. 74180; Direct Blue 86
DKBXPLYSDKSFEQ-UHFFFAOYSA-L
780.16
Odorless

Chemical Identifiers

1328-51-4
780.16
DKBXPLYSDKSFEQ-UHFFFAOYSA-L
92030797
C32H14CuN8Na2O6S2
MFCD00071424
C.I. 74180; Direct Blue 86
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-523-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only