missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Solvent Blue 38

Catalog No. p-7022559
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1539514 25 g
AAA1539522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1539514 Supplier Thermo Scientific Chemicals Supplier No. A1539514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol

Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.

Solubility
Soluble in water (partly), and methanol (1 mg/ml).

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1328-51-4
Molecular Formula C32H14CuN8Na2O6S2
Molecular Weight (g/mol) 780.16
MDL Number MFCD00071424
InChI Key DKBXPLYSDKSFEQ-UHFFFAOYSA-L
PubChem CID 92030797
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2]

Specifications

Melting Point ∼235°C (decomposition)
Solubility Information Soluble in water (partly),and methanol (1mg/ml).
Quantity 25 g
Chemical Name or Material Fast Blue
EINECSNumber 215-523-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only