Learn More
Thermo Scientific Chemicals Solvent Blue 38
CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol
Supplier: Thermo Scientific Chemicals A1539514
Description
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSolvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.
Solubility
Soluble in water (partly), and methanol (1 mg/ml).
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
| Fast Blue | |
| 1328-51-4 | |
| MFCD00071424 | |
| DKBXPLYSDKSFEQ-UHFFFAOYSA-L | |
| 780.16 | |
| 25 g |
| ∼235°C (decomposition) | |
| C32H14CuN8Na2O6S2 | |
| Soluble in water (partly),and methanol (1mg/ml). | |
| C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2] | |
| 92030797 |
Chemical Identifiers
| 1328-51-4 | |
| 780.16 | |
| DKBXPLYSDKSFEQ-UHFFFAOYSA-L | |
| C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2] |
| C32H14CuN8Na2O6S2 | |
| MFCD00071424 | |
| 92030797 |
Safety and Handling
EINECSNumber : 215-523-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only