missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solvent blue 35, MP Biomedicals™
$680.74 - $680.74
Chemical Identifiers
| CAS | 17354-14-2 |
|---|---|
| Molecular Formula | C22H26N2O2 |
| Molecular Weight (g/mol) | 350.462 |
| MDL Number | MFCD00011714 |
| InChI Key | OCQDPIXQTSYZJL-UHFFFAOYSA-N |
| Synonym | C.I. 61554, Sudan Blue II |
| PubChem CID | 3766139 |
| IUPAC Name | 1,4-bis(butylamino)anthracene-9,10-dione |
| SMILES | CCCCNC1=C2C(=C(C=C1)NCCCC)C(=O)C3=CC=CC=C3C2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15663580
|
MP Biomedicals Inc
0215663580 |
100 g |
Each for $680.74
|
|
|||||
Chemical Identifiers
| 17354-14-2 | |
| 350.462 | |
| OCQDPIXQTSYZJL-UHFFFAOYSA-N | |
| 3766139 | |
| CCCCNC1=C2C(=C(C=C1)NCCCC)C(=O)C3=CC=CC=C3C2=O |
| C22H26N2O2 | |
| MFCD00011714 | |
| C.I. 61554, Sudan Blue II | |
| 1,4-bis(butylamino)anthracene-9,10-dione |
Specifications
| 17354-14-2 | |
| MFCD00011714 | |
| OCQDPIXQTSYZJL-UHFFFAOYSA-N | |
| 1,4-bis(butylamino)anthracene-9,10-dione | |
| 3766139 | |
| 1.23g/mL | |
| Powder |
| C22H26N2O2 | |
| C.I. 61554, Sudan Blue II | |
| CCCCNC1=C2C(=C(C=C1)NCCCC)C(=O)C3=CC=CC=C3C2=O | |
| 350.462 | |
| 350.5 | |
| 100 g | |
| Solvent Blue 35, Dye content: Approx. 90% |
Safety and Handling
EINECSNumber : 241-379-4
Recommended Storage : Store at Room Temperature (15-30°C).