missing translation for 'onlineSavingsMsg'
Learn More

Solvent blue 35, MP Biomedicals™

Catalog No. icn15663580
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15663580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15663580 Supplier MP Biomedicals Inc Supplier No. 0215663580
Only null left

Chemical Identifiers

CAS 17354-14-2
Molecular Formula C22H26N2O2
Molecular Weight (g/mol) 350.462
MDL Number MFCD00011714
InChI Key OCQDPIXQTSYZJL-UHFFFAOYSA-N
Synonym C.I. 61554, Sudan Blue II
PubChem CID 3766139
IUPAC Name 1,4-bis(butylamino)anthracene-9,10-dione
SMILES CCCCNC1=C2C(=C(C=C1)NCCCC)C(=O)C3=CC=CC=C3C2=O

Specifications

Formula Weight 350.5
Density 1.23g/mL
Quantity 100 g
Physical Form Powder
Chemical Name or Material Solvent Blue 35, Dye content: Approx. 90%
EINECSNumber 241-379-4
Recommended Storage Store at Room Temperature (15-30°C).