missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Solus™ 2000 Series Protective Eyewear
Comfortable, stylish eyewear with a vast array of color and lens tint options. Eye protection with a stylish flare.
Supplier: 3M™ 7100203166
Description
- Adjustable ratchet temples
- Polycarbonate lenses absorb 99.9% of UVA and UVB rays
- Meet the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
- 3M Catalog# S2001AF-BLU, S2001AS-BLU, S2001SGAF-BGR, S2001SGAF-BGR-F, S2001SGAF-RED, S2002SGAF-BGR, S2002SGAF-RED, S2003SGAF-BGR, S2005SGAF-BGR, S2007SGAF-BGR, S2017AS-BLK, S2024AS-RED, S2030AS-BLK, S2050AS-BLK
Specifications
| Anti-fog, Scratch-resistant | |
| Protective Eyewear | |
| Gray | |
| Dual Injected Temple | |
| Polycarbonate | |
| UV400 |
| Brown | |
| Assembly, Law Enforcement, Primary Metals, Power Fastening, Pouring/Casting | |
| S2005SGAF-BGR | |
| Gray, Blue-Green | |
| Plastic |