Learn More
3M™ Solus™ 2000 Series Protective Eyewear
Catalog No. p-7163269
Change view
Click to view available options
Lens Tint:
Brown
Clear
Lens Coating:
Anti-fog, Scratch-resistant
Temple Color:
Gray, Blue-Green
Model:
S2001SGAF-BGR
S2001SGAF-BGR-F
S2005SGAF-BGR
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating Temple Color Model
19071177 Clear Anti-fog, Scratch-resistant Gray, Blue-Green S2001SGAF-BGR
19071181 Brown Anti-fog, Scratch-resistant Gray, Blue-Green S2005SGAF-BGR
19071184 Clear Anti-fog, Scratch-resistant Gray, Blue-Green S2001SGAF-BGR-F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19071177 Supplier 3M™ Supplier No. 7100203178
Only null left
Comfortable, stylish eyewear with a vast array of color and lens tint options. Eye protection with a stylish flare.
  • Adjustable ratchet temples
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays
  • Meet the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
  • 3M Catalog# S2001AF-BLU, S2001AS-BLU, S2001SGAF-BGR, S2001SGAF-BGR-F, S2001SGAF-RED, S2002SGAF-BGR, S2002SGAF-RED, S2003SGAF-BGR, S2005SGAF-BGR, S2007SGAF-BGR, S2017AS-BLK, S2024AS-RED, S2030AS-BLK, S2050AS-BLK
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Protective Eyewear
For Use With (Application) Assembly, Law Enforcement, Primary Metals, Power Fastening, Pouring/Casting
Frame Color Gray
Model S2001SGAF-BGR
Temple Style Dual Injected Temple
Temple Color Gray, Blue-Green
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Show More Show Less