missing translation for 'onlineSavingsMsg'
Learn More

Solketal, 97%

CAS: 100-79-8 | C6H12O3 | 132.159 g/mol

Supplier:  Thermo Scientific Chemicals L0281436

 View more versions of this product

Catalog No. AAL0281436


Only null left
Explore available promotions

Description

Description

Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .

Solubility
Soluble in water (fully), alcohol, acetals, esters, and ethers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are acids, Strong oxidizing agents.
Specifications

Specifications

Solketal
-27°C
188°C to 190°C
C6H12O3
MFCD00063238
104465
solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
RNVYQYLELCKWAN-UHFFFAOYSA-N
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
7528
97%
100-79-8
1.064
80°C (176°F)
1.434
500 g
14,5213
Soluble in water (fully),alcohol,acetals,esters,and ethers.
CC1(OCC(O1)CO)C
132.159
132.16

Chemical Identifiers

100-79-8
132.159
RNVYQYLELCKWAN-UHFFFAOYSA-N
7528
CC1(OCC(O1)CO)C
C6H12O3
MFCD00063238
solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P264b-P280-P305+P351+P338-P370+P378q-P501c

H227-H319

EINECSNumber : 202-888-7

RTECSNumber : JI0400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only