missing translation for 'onlineSavingsMsg'
Learn More

Solketal, 97%

Catalog No. p-7043804
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0281422 100 g
AAL0281436 500 g
AAL028140E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0281422 Supplier Thermo Scientific Chemicals Supplier No. L0281422
Only null left

CAS: 100-79-8 | C6H12O3 | 132.159 g/mol

Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .

Solubility
Soluble in water (fully), alcohol, acetals, esters, and ethers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are acids, Strong oxidizing agents.

Chemical Identifiers

CAS 100-79-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00063238
InChI Key RNVYQYLELCKWAN-UHFFFAOYSA-N
Synonym solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
PubChem CID 7528
IUPAC Name (2,2-dimethyl-1,3-dioxolan-4-yl)methanol
SMILES CC1(OCC(O1)CO)C

Specifications

Melting Point -27°C
Density 1.064
Boiling Point 188°C to 190°C
Flash Point 80°C (176°F)
Refractive Index 1.434
Quantity 100 g
Beilstein 104465
Merck Index 14,5213
Solubility Information Soluble in water (fully),alcohol,acetals,esters,and ethers.
Formula Weight 132.16
Percent Purity 97%
Chemical Name or Material Solketal
Show More Show Less
Hazard Category H227-H319
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 202-888-7
RTECSNumber JI0400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only