Learn More
MilliporeSigma™ Supelco™ Solid Top Screw Cap with PTFE Liner
Known for providing the widest range of chemical compatibility and for being the most temperature tolerant of plastic closures
$43.60 - $97.20
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|
111020924
|
MilliporeSigma™ Supelco™
27141 |
4 mL Vial |
Each for $96.40
|
|
|||||
|
111011425
|
MilliporeSigma™ Supelco™
27091U |
2 mL Standard Opening Vial |
Each for $97.20
|
|
|||||
|
111011936
|
MilliporeSigma™ Supelco™
28659U |
2 mL Large Opening Vial |
Each for $43.60
|
|
|||||
Description
- The color green easily identifies the PTFE liner system. F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature and provides odor and impact resilience for a tight seal with a low moisture transmission rate.
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene).
- Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Specifications
| F217/PTFE | |
| 100 |