Learn More
Fisherbrand™ Solid Rubber Stoppers
Available in a variety of sizes ranging from 000 through 16
 View more versions of this product
Catalog No. 14-130P
Only null left
Description
Description
Available in a variety of sizes ranging from 000 through 16
  • Solid rubber construction
The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).
Specifications
Specifications
Black
10
10
5 cm
2.5 cm
Solid Stopper
General purpose use
4.2 cm
Natural Rubber
Product Suggestions
Product Suggestions