missing translation for 'onlineSavingsMsg'
Learn More

Sodium tungstate dihydrate, 99+%, ACS reagent

Catalog No. AC424470050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424470050 5 g Glass bottle
AC424471000 100 g Glass bottle
AC424475000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424470050 Supplier Thermo Scientific Chemicals Supplier No. 424470050

CAS: 10213-10-2 | Na2O42H2O | 329.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 10213-10-2
Molecular Formula Na2O42H2O
Molecular Weight (g/mol) 329.85
MDL Number MFCD00149190
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
PubChem CID 150191
ChEBI CHEBI:63939
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White to Yellow
Physical Form Slightly Crystalline Powder or Flakes
Quantity 5 g
Assay Percent Range 99+%
Linear Formula Na2WO4·2H2O
Fieser 15,295
Merck Index 15, 8826
Solubility Information Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol and acids
Formula Weight 329.85
Percent Purity ≥99%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Sodium tungstate dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

RUO – Research Use Only