missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tripolyphosphate, Anhydrous, FCC, 85%, Spectrum™ Chemical

Catalog No. 18606901
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606901 2.5 kg Poly Pail
18606902 12 kg Poly Pail
18606903 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606901 Supplier Spectrum Chemical Mfg Cor Supplier No. S15082.5KG

S1508, 7758-29-4, Na5P3O10

Spectrum™ Chemical Sodium Tripolyphosphate, Anhydrous, FCC is a food preservative and as an emulsifier to retain moisture. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7758-29-4
Molecular Formula Na5O10P3
Molecular Weight (g/mol) 367.86
InChI Key HWGNBUXHKFFFIH-UHFFFAOYSA-I
IUPAC Name pentasodium bis(phosphonatooxy)phosphinate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 85%
Grade FCC
Packaging Poly Pail