accessibility menu, dialog, popup

UserName

NaNaNaNaNaOPOOOPOOOPOOO

CAS RN 7758-29-4

IUPAC Name: pentasodium bis(phosphonatooxy)phosphinate
Synonyms: sodium triphosphate
Molecular Weight (g/mol): 367.86
Molecular Formula: Na5O10P3
InChi Key: HWGNBUXHKFFFIH-UHFFFAOYSA-I
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O
Grade 
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (7)
Physical Form 
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (4)
  • (7)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (5)
  • (7)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (6)