missing translation for 'onlineSavingsMsg'
Learn More

Sodium tripolyphosphate, 85%, pure, Tech.

CAS: 7758-29-4 | Na5O10P3 | 367.85 g/mol

Supplier:  Thermo Scientific Chemicals 218670025

 View more versions of this product

Catalog No. AC218670025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium tripolyphosphate
tech., 85%
White
2.5 kg
Na5O10P3
MFCD00003514
sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n
HWGNBUXHKFFFIH-UHFFFAOYSA-I
pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate
24455
85%
Plastic bottle
622.0°C
7758-29-4
Crystalline Powder
85%
Na5P3O10
15, 8824
Solubility in water: 145g/L (20°C)
[O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]
367.85
367.85
Pure

Chemical Identifiers

7758-29-4
367.85
HWGNBUXHKFFFIH-UHFFFAOYSA-I
24455
[O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]
Na5O10P3
MFCD00003514
sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n
pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 231-838-7

RTECSNumber : YK4570000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only