missing translation for 'onlineSavingsMsg'
Learn More

Sodium trimethylsilanolate, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog No. p-4089858
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428841000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428841000 Supplier Thermo Scientific Chemicals Supplier No. 428841000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18027-10-6 , 75-09-2
Molecular Formula C3H9NaOSi
Molecular Weight (g/mol) 112.18
MDL Number MFCD00013926
InChI Key HSNUIYJWTSJUMS-UHFFFAOYSA-N
Synonym sodium trimethylsilanolate, silanol, trimethyl-, sodium salt, trimethylsilanol sodium salt, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol,1,1,1-trimethyl-, sodium salt 1:1, naotms, sodiooxytrimethylsilane, naosi me 3, trimethylsilyloxy sodium, sodium trimethylsilanolat
PubChem CID 23672319
IUPAC Name sodium;trimethyl(oxido)silane
SMILES C[Si](C)(C)[O-].[Na+]

Specifications

Quantity 100 mL
Flash Point 63°C
Formula Weight 112.18
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.28
Density 1.2800g/mL
Chemical Name or Material Sodium trimethylsilanolate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
May cause drowsiness or dizziness.<b
Precautionary Statement GHS P Statement
Do not handle until all safety precautions have been read and understood.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pres
EINECSNumber 241-939-8

RUO – Research Use Only