missing translation for 'onlineSavingsMsg'
Learn More

Sodium (Trihydroxy)phenylborate 98.0+%, TCI America™

Catalog No. S05585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05585G Supplier TCI America Supplier No. S05585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52542-80-0
Molecular Formula C6H8BNaO3
Molecular Weight (g/mol) 161.927
InChI Key ZNXSFEXRXNJDNK-UHFFFAOYSA-N
PubChem CID 44629933
IUPAC Name sodium;trihydroxy(phenyl)boranuide
SMILES [B-](C1=CC=CC=C1)(O)(O)O.[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 161.93
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium (Trihydroxy)phenylborate