accessibility menu, dialog, popup

UserName

BOHOHHONa

CAS RN 52542-80-0

IUPAC Name: sodium trihydroxy(phenyl)boranuide
Synonyms: sodium trihydroxy(phenyl)boranuide
Molecular Weight (g/mol): 161.93
Molecular Formula: C6H8BNaO3
InChi Key: ZNXSFEXRXNJDNK-UHFFFAOYSA-N
SMILES: [Na+].O[B-](O)(O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results