missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Trifluoromethanesulfonate 98.0+%, TCI America™
Supplier: TCI America T155025G
Specifications
| Sodium Trifluoromethanesulfonate | |
| White | |
| MFCD00061607 | |
| sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d | |
| C(F)(F)(F)S(=O)(=O)[O-].[Na+] | |
| 172.053 | |
| 172.05 | |
| Crystalline Powder |
| 2926-30-9 | |
| CF3NaO3S | |
| 25 g | |
| XGPOMXSYOKFBHS-UHFFFAOYSA-M | |
| sodium;trifluoromethanesulfonate | |
| 23676748 | |
| ≥98.0% (T) |
Chemical Identifiers
| 2926-30-9 | |
| 172.053 | |
| XGPOMXSYOKFBHS-UHFFFAOYSA-M | |
| 23676748 | |
| C(F)(F)(F)S(=O)(=O)[O-].[Na+] |
| CF3NaO3S | |
| MFCD00061607 | |
| sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d | |
| sodium;trifluoromethanesulfonate |
Safety and Handling
TSCA : No