missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium trifluoromethanesulfonate, 98%
Description
Sodium trifluoromethanesulfonate is used in the preparation of N-fluoro-2-methylpyridinium triflate by reaction with dinitrogen difluoride as a reagent. It is also used as a chaotropic mobile phase additive in reversed-phase liquid chromatography (RP-LC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium trifluoromethanesulfonate is used in the preparation of N-fluoro-2-methylpyridinium triflate by reaction with dinitrogen difluoride as a reagent. It is also used as a chaotropic mobile phase additive in reversed-phase liquid chromatography (RP-LC).
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids and bases.
Sodium trifluoromethanesulfonate is used in the preparation of N-fluoro-2-methylpyridinium triflate by reaction with dinitrogen difluoride as a reagent. It is also used as a chaotropic mobile phase additive in reversed-phase liquid chromatography (RP-LC).
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids and bases.
Specifications
Specifications
| Melting Point | 251°C to 255°C |
| Quantity | 5 g |
| Beilstein | 3728797 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 172.06 |
| Percent Purity | 98% |
| Chemical Name or Material | Sodium trifluoromethanesulfonate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only