missing translation for 'onlineSavingsMsg'
Learn More

Sodium Trifluoromethanesulfinate 95.0+%, TCI America™

Catalog No. T20335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20335G 5 g
T203325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20335G Supplier TCI America Supplier No. T20335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2926-29-6
Molecular Formula CF3NaO2S
Molecular Weight (g/mol) 156.054
MDL Number MFCD03092989
InChI Key KAVUKAXLXGRUCD-UHFFFAOYSA-M
Synonym sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
PubChem CID 23690734
IUPAC Name sodium;trifluoromethanesulfinate
SMILES C(F)(F)(F)S(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 156.05
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Trifluoromethanesulfinate