accessibility menu, dialog, popup

UserName

FFFSOONa

CAS RN 2926-29-6

IUPAC Name: sodium trifluoromethanesulfinate
Synonyms: sodium trifluoromethanesulfinate
Molecular Weight (g/mol): 156.05
Molecular Formula: CF3NaO2S
InChi Key: KAVUKAXLXGRUCD-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)