missing translation for 'onlineSavingsMsg'
Learn More

Sodium trifluoromethanesulfinate, 98%

Catalog No. p-7077526
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454990010 1 g
AC454990050 5 g
AC454990250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC454990010 Supplier Thermo Scientific Chemicals Supplier No. 454990010

CAS: 2926-29-6 | CF3NaO2S | 156.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2926-29-6
Molecular Formula CF3NaO2S
Molecular Weight (g/mol) 156.06
InChI Key KAVUKAXLXGRUCD-UHFFFAOYSA-M
Synonym sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
PubChem CID 23690734
IUPAC Name sodium;trifluoromethanesulfinate
SMILES C(F)(F)(F)S(=O)[O-].[Na+]

Specifications

Melting Point 350.0°C
Quantity 1 g
Assay Percent Range 98%
Linear Formula CF3SO2Na
Solubility Information Solubility in water: soluble.
Formula Weight 156.06
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Sodium trifluoromethanesulfinate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only