missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tricyanomethanide 98.0+%, TCI America™
Supplier: TCI America O04895G
Specifications
| Sodium Tricyanomethanide | |
| C4N3Na | |
| 3439 | |
| LOYHZHALCDOXIK-UHFFFAOYSA-N | |
| sodium;methanetricarbonitrile | |
| 24840070 | |
| ≥98.0% (T) |
| 36603-80-2 | |
| 5 g | |
| Tricyanomethanide Sodium Salt | |
| C(#N)[C-](C#N)C#N.[Na+] | |
| 113.055 | |
| 113.05 | |
| Crystalline Powder |
Chemical Identifiers
| 36603-80-2 | |
| 113.055 | |
| Tricyanomethanide Sodium Salt | |
| sodium;methanetricarbonitrile |
| C4N3Na | |
| LOYHZHALCDOXIK-UHFFFAOYSA-N | |
| 24840070 | |
| C(#N)[C-](C#N)C#N.[Na+] |
Safety and Handling
TSCA : No