missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tricyanomethanide 98.0+%, TCI America™

Catalog No. p-7134360
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04895G 5 g
O048925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04895G Supplier TCI America Supplier No. O04895G
Only null left

Chemical Identifiers

CAS 36603-80-2
Molecular Formula C4N3Na
Molecular Weight (g/mol) 113.055
InChI Key LOYHZHALCDOXIK-UHFFFAOYSA-N
Synonym Tricyanomethanide Sodium Salt
PubChem CID 24840070
IUPAC Name sodium;methanetricarbonitrile
SMILES C(#N)[C-](C#N)C#N.[Na+]

Specifications

Quantity 5 g
UN Number 3439
Formula Weight 113.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Tricyanomethanide