missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Triacetoxyborohydride 80.0+%, TCI America™
Supplier: TCI America S0394100G
Specifications
| Sodium Triacetoxyborohydride | |
| 106°C | |
| C6H10BNaO6 | |
| 100 g | |
| c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate | |
| [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O | |
| 211.94 | |
| ≥80.0% (T) |
| 56553-60-7 | |
| White | |
| MFCD00012211 | |
| 2813 | |
| HHYFEYBWNZJVFQ-UHFFFAOYSA-N | |
| sodium bis(acetyloxy)boranuidyl acetate | |
| 211.94 | |
| Crystalline Powder |
Chemical Identifiers
| 56553-60-7 | |
| 211.94 | |
| HHYFEYBWNZJVFQ-UHFFFAOYSA-N | |
| sodium bis(acetyloxy)boranuidyl acetate |
| C6H10BNaO6 | |
| MFCD00012211 | |
| c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate | |
| [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O |
Safety and Handling
EINECSNumber : (2)-4147
TSCA : Yes