missing translation for 'onlineSavingsMsg'
Learn More

Sodium Triacetoxyborohydride 80.0+%, TCI America™

Catalog No. p-7135876
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S039425G 25 g
S0394100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S039425G Supplier TCI America Supplier No. S039425G
Only null left

Chemical Identifiers

CAS 56553-60-7
Molecular Formula C6H10BNaO6
Molecular Weight (g/mol) 211.94
MDL Number MFCD00012211
InChI Key HHYFEYBWNZJVFQ-UHFFFAOYSA-N
Synonym c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
IUPAC Name sodium bis(acetyloxy)boranuidyl acetate
SMILES [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O

Specifications

Melting Point 106°C
Color White
Quantity 25 g
UN Number 2813
Formula Weight 211.94
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Triacetoxyborohydride
EINECSNumber (2)-4147
TSCA Yes