missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate solution, Volumetric, Reag. Ph. Eur., 0.1Â M Na2S2O3 (0.1N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.1Â M Na2S2O3 (0.1N)
$132.53 - $707.68
Chemical Identifiers
| CAS | 7772-98-7 |
|---|---|
| Molecular Formula | Na2O3S2 |
| Molecular Weight (g/mol) | 158.10 |
| MDL Number | MFCD00003499 |
| InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
| PubChem CID | 24477 |
| SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6002621
|
Honeywell-Fluka
352451L |
1 L |
Each for $132.53
|
|
|||||
|
6002622
|
Honeywell-Fluka
352455LVP |
5 L |
Each for $403.78
|
|
|||||
|
60046547
|
Honeywell-Fluka
352456X1L |
6 x 1 L |
Case of 6 Each for $633.55
|
|
|||||
|
6002620
|
Honeywell-Fluka
3524510LVP |
10 L |
Each for $707.68
|
|
|||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
| 7772-98-7 | |
| 158.10 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| 24477 |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| [Na+].[Na+].[O-]S([S-])(=O)=O |
Specifications
| 7772-98-7 | |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| [Na+].[Na+].[O-]S([S-])(=O)=O | |
| 158.10 | |
| 158.11g/mol |
| 1 L | |
| Na2S2O3 | |
| NONH for all modes of transport | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
| 24477 | |
| Sodium thiosulfate solution |
Safety and Handling
ShelfLife : 1080 days from date of manufacture
EINECSNumber : 231-791-2
Recommended Storage : Room Temp