missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Solution, 0.0394M, Honeywell Fluka™
Volumetric, 0.0394Â M Na2S2O3 (0.0394N), for the determination of iodine adsorption number according to ASTM D 1510
$275.00 - $275.00
Chemical Identifiers
| CAS | 7772-98-7 |
|---|---|
| Molecular Formula | Na2O3S2 |
| Molecular Weight (g/mol) | 158.10 |
| MDL Number | MFCD00003499 |
| InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
| PubChem CID | 24477 |
| SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
| 7772-98-7 | |
| 158.10 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| 24477 |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| [Na+].[Na+].[O-]S([S-])(=O)=O |
Specifications
| 7772-98-7 | |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| [Na+].[Na+].[O-]S([S-])(=O)=O | |
| 158.10 | |
| 158.11g/mol | |
| Sodium thiosulfate solution |
| 1 L | |
| Na2S2O3 | |
| NONH for all modes of transport | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
| 24477 | |
| 1.01g/mL at 20°C |