missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Catalog No. p-7072035
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
Catalog No. Quantity
6002377 1 kg
60045959 6 x 1 kg
2 options

Catalog No. 6002377

Supplier: Honeywell Chemicals 314591KG

Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Quantity 1 kg
Linear Formula Na2S2O3 · 5H2O
MDL Number MFCD00149186
UN Number NONH for all modes of transport
Formula Weight 248.18g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Thiosulfate Pentahydrate, Reag. Ph. Eur. reag. ISO
EINECSNumber 231-867-5