missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Pentahydrate, puriss., Honeywell Fluka™

Catalog No. p-7071814
Change view
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002000 500 g
6001999 2.5 kg
60044815 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6002000 Supplier Honeywell Chemicals Supplier No. 13479500G
Only null left

Puriss., meets analytical specification of BP, European Pharmacopoeia, USP, 99-101%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
MDL Number MFCD00149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Quantity 500 g
Linear Formula Na2S2O3 · 5H2O
UN Number NONH for all modes of transport
Formula Weight 248.18g/mol
Percent Purity 99 to 101%
Grade puriss.
Chemical Name or Material Sodium Thiosulfate Pentahydrate, Meets analytical specification of Ph. Eur.BP, USP
EINECSNumber 231-867-5