missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Pentahydrate, ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company RDCS0930500B1
| Quantity | 500 g |
|---|---|
| Packaging | White Poly Wide Mouth Bottle |
Chemical Identifiers
| 10102-17-7 , 497-19-8 | |
| 248.172 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 | |
| O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
Specifications
| Sodium Thiosulfate | |
| 10102-17-7 , 497-19-8 | |
| 100.0, 0.04, 3.0E-4 | |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
| 61475 | |
| ACS |
| Pentahydrate | |
| 98.0, 0.04, 3.0E-4 | |
| 500 g | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
| 248.172 | |
| CHEBI:32150 | |
| White Poly Wide Mouth Bottle |
Safety and Handling
ShelfLife : 60 months