missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, ACS, 99.5-101.0%

Catalog No. p-4407947
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1451836 500 g
AA14518A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1451836 Supplier Thermo Scientific Chemicals Supplier No. 01451836
Only null left

Maximum level of impurities: Insoluble matter 0.005%, pH of a 5% solution 6.0-8.4 at 25°, Nitrogen compounds (as N) 0.002%, Sulfate and sulfite (as SO4) 0.1%, Sulfide P.T. (limit about 1ppm) | CAS: 10102-17-7 | H10Na2O8S2 | 248.172 g/mol

Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivatives, S-alkylthiosulfonates, are known as Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solutions and to check the pH of bleach solutions with liquid indicators. Other uses include tanning of leather; as an active component in hand warmers and chemical heating pads and as an anionic surfactant to aid in the dispersion of pharmaceutical preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
MDL Number MFCD00149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Boiling Point 100°C-5H{2}O
Melting Point 48°C
Color Colorless
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.5-101.0%
Solubility Information Soluble in water and ammonia. Insoluble in alcohol and organic solvents.
Formula Weight 248.17 (158.10 Anhydrous)
Grade ACS
Odor Odorless
Appearance Colorless crystalline
Density 1.69 g/cm3
Chemical Name or Material Sodium thiosulfate pentahydrate, For ACS analysis
Show More Show Less
Hazard Category H500
EINECSNumber 231-867-5
RTECSNumber WE6660000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only